Learn More
Medchemexpress LLC Mal-PEG5-NHS ester | 1807537-42-3 | 99.0% | 50 MG
SDP
Catalog No. 5000341584
Encompass_Preferred
Mal-PEG5-NHS ester is an alkyl/ether and PEG-based PROTAC linker. It is utilized in the synthesis of Proteolysis-targeting chimeras (PROTACs), which are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether and PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Designed to selectively degrade target proteins
Catalog No. 50-003-41584 Supplier Medchemexpress LLC Supplier No. HY12688850MG
Mal-PEG5-NHS ester is an alkyl/ether and PEG-based PROTAC linker. It is utilized in the synthesis of Proteolysis-targeting chimeras (PROTACs), which are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether and PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Designed to selectively degrade target proteins