Learn More
Medchemexpress LLC Mal-PEG5-NHS ester | 1807537-42-3 | 99.04% | 25 MG
SDP
Catalog No. 5000342698
Encompass_Preferred
Mal-PEG5-NHS ester is an Alkyl/ether and PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • Alkyl/ether and PEG-based PROTAC linker
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Contains two different ligands connected by a linker
Catalog No. 50-003-42698 Supplier Medchemexpress LLC Supplier No. 25MGHY126888
Mal-PEG5-NHS ester is an Alkyl/ether and PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • Alkyl/ether and PEG-based PROTAC linker
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Contains two different ligands connected by a linker