Learn More
Medchemexpress LLC Mal-PEG6-acid | 518044-42-3 | ≥98.0% | 250 MG
SDP
Catalog No. 5000343022
Encompass_Preferred
Mal-PEG6-acid is a polyethylene glycol-based linker employed in the creation of protein degraders.
  • It is a PEG-based linker.
  • It can be used in the synthesis of protein degraders.
  • It has high purity (≥98.0%).
Catalog No. 50-003-43022 Supplier Medchemexpress LLC Supplier No. 250MGHY126963
Mal-PEG6-acid is a polyethylene glycol-based linker employed in the creation of protein degraders.
  • It is a PEG-based linker.
  • It can be used in the synthesis of protein degraders.
  • It has high purity (≥98.0%).