Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG6-NHS ester | 1599472-25-9 | MFCD20926374 | 98.0% | 530.52 g/mol | C23H34N2O12 | 1 G
SDP

Catalog No. 5000202612
Encompass_Preferred

Mal-PEG6-NHS ester is a non-cleavable ADC linker containing a maleimide reactive group, a six-unit polyethylene glycol (PEG6) spacer, and an N-hydroxysuccinimide (NHS) ester for amine coupling. It is supplied for research use in synthesis and bioconjugation workflows.

  • Purity 98.0%.
  • Cas number 1599472-25-9.
  • Molecular weight 530.52 g/mol.
  • Chemical formula C23H34N2O12.
  • Soluble in DMSO at 100 mg/mL (in vitro; ultrasonic recommended).
  • Suggested in vivo formulations provided for dosing preparations.
  • For research use only; not for use in patients.
  • Pack quantity 1 G.

Catalog No. 50-002-02612 Supplier Medchemexpress LLC Supplier No. HY1300851G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG6-NHS ester is a non-cleavable ADC linker containing a maleimide reactive group, a six-unit polyethylene glycol (PEG6) spacer, and an N-hydroxysuccinimide (NHS) ester for amine coupling. It is supplied for research use in synthesis and bioconjugation workflows.

  • Purity 98.0%.
  • Cas number 1599472-25-9.
  • Molecular weight 530.52 g/mol.
  • Chemical formula C23H34N2O12.
  • Soluble in DMSO at 100 mg/mL (in vitro; ultrasonic recommended).
  • Suggested in vivo formulations provided for dosing preparations.
  • For research use only; not for use in patients.
  • Pack quantity 1 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.