Learn More
Medchemexpress LLC Mal-PEG8-NHS ester | 2055033-05-9 | 98.96% | 25 MG
SDP
Catalog No. 5000342674
Encompass_Preferred
Mal-PEG8-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This compound is for research use only and aids in the development of PROTACs, which exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-42674 Supplier Medchemexpress LLC Supplier No. 25MGHY126887
Mal-PEG8-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This compound is for research use only and aids in the development of PROTACs, which exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only