Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG8-NHS ester | 2055033-05-9 | 99.0% | 1 G
SDP

Catalog No. 5000341545
Encompass_Preferred

Mal-PEG8-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight: 618.63
  • Molecular formula: C27H42N2O14
  • Purity: 98.96%
  • Appearance: Colorless to light yellow viscous liquid
  • Soluble in DMSO: ≥ 100 mg/mL

Catalog No. 50-003-41545 Supplier Medchemexpress LLC Supplier No. HY1268871G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-PEG8-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight: 618.63
  • Molecular formula: C27H42N2O14
  • Purity: 98.96%
  • Appearance: Colorless to light yellow viscous liquid
  • Soluble in DMSO: ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.