Learn More
Medchemexpress LLC Mal-PEG8-NHS ester | 2055033-05-9 | 99.0% | 100 MG
SDP
Catalog No. 5000340830
Encompass_Preferred
Mal-PEG8-NHS ester is a PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. PROTACs are innovative molecules designed to exploit the ubiquitin-proteasome system, selectively degrading target proteins by linking an E3 ubiquitin ligase ligand to a target protein ligand.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Designed to selectively degrade target proteins
  • For research use only
Catalog No. 50-003-40830 Supplier Medchemexpress LLC Supplier No. HY126887100MG
Mal-PEG8-NHS ester is a PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. PROTACs are innovative molecules designed to exploit the ubiquitin-proteasome system, selectively degrading target proteins by linking an E3 ubiquitin ligase ligand to a target protein ligand.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Designed to selectively degrade target proteins
  • For research use only