Learn More
Medchemexpress LLC Mal-PEG8-NHS ester | 2055033-05-9 | 99.0% | 250 MG
SDP
Catalog No. 5000341983
Encompass_Preferred
Mal-PEG8-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) are molecules that harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Consists of two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • Other ligand for the target protein
Catalog No. 50-003-41983 Supplier Medchemexpress LLC Supplier No. 250MGHY126887
Mal-PEG8-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs (Proteolysis Targeting Chimeras) are molecules that harness the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Consists of two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • Other ligand for the target protein