Learn More
Medchemexpress LLC Mal-PEG8-Phe-Lys-PAB-Exatecan TFA | 2679821-44-2 | 98.7% | 1434.56 g/mol | C73H92FN9O20 | 10 MG
SDP
Catalog No. 5000287395
Encompass_Preferred
Mal-PEG8-Phe-Lys-PAB-Exatecan TFA is a drug-linker conjugate for antibody-drug conjugate (ADC) research that links a cleavable Phe-Lys-PAB linker and an 8-unit PEG spacer to the camptothecin-derived toxin exatecan, supplied as the trifluoroacetate (TFA) salt. The reagent is provided as a solid in small research pack sizes and is intended for conjugation chemistry, payload evaluation, and linker optimization in preclinical workflows.
  • Contains a cleavable Phe-Lys-PAB linker for protease-triggered payload release.
  • Includes a PEG8 spacer to improve solubility and reduce aggregation.
  • Features a maleimide group for thiol-selective conjugation.
  • Payload is a camptothecin-derived exatecan presented as a TFA salt.
  • High purity suitable for research applications.
  • Provided in small pack sizes for preclinical and chemical biology use.
Catalog No. 50-002-87395 Supplier Medchemexpress LLC Supplier No. HY148380A10MG
Mal-PEG8-Phe-Lys-PAB-Exatecan TFA is a drug-linker conjugate for antibody-drug conjugate (ADC) research that links a cleavable Phe-Lys-PAB linker and an 8-unit PEG spacer to the camptothecin-derived toxin exatecan, supplied as the trifluoroacetate (TFA) salt. The reagent is provided as a solid in small research pack sizes and is intended for conjugation chemistry, payload evaluation, and linker optimization in preclinical workflows.
  • Contains a cleavable Phe-Lys-PAB linker for protease-triggered payload release.
  • Includes a PEG8 spacer to improve solubility and reduce aggregation.
  • Features a maleimide group for thiol-selective conjugation.
  • Payload is a camptothecin-derived exatecan presented as a TFA salt.
  • High purity suitable for research applications.
  • Provided in small pack sizes for preclinical and chemical biology use.