Learn More
Medchemexpress LLC Mal-PEG8-Phe-Lys-PAB-Exatecan TFA | 2679821-44-2 | 98.7% | 1548.58 | 1 MG
SDP
Catalog No. 5000360342
Encompass_Preferred
Mal-PEG8-Phe-Lys-PAB-Exatecan TFA is an agent-linker conjugate for ADC, composed of the degradable linker Mal-PEG8-Phe-Lys-PAB and the toxin molecule Exatecan. It is for research use only.
  • Agent-linker conjugate for ADCs
  • Carries a highly potent small-molecule toxin
  • Targets specific antigens on tumor cell surfaces with high binding affinity
  • Internalized into tumor cells and digested to release antitumor toxin
  • Appears as a light yellow to yellow solid
Catalog No. 50-003-60342 Supplier Medchemexpress LLC Supplier No. HY148380A1MG
Mal-PEG8-Phe-Lys-PAB-Exatecan TFA is an agent-linker conjugate for ADC, composed of the degradable linker Mal-PEG8-Phe-Lys-PAB and the toxin molecule Exatecan. It is for research use only.
  • Agent-linker conjugate for ADCs
  • Carries a highly potent small-molecule toxin
  • Targets specific antigens on tumor cell surfaces with high binding affinity
  • Internalized into tumor cells and digested to release antitumor toxin
  • Appears as a light yellow to yellow solid