Learn More
Medchemexpress LLC Mal-PEG8-Phe-Lys-PAB-Exatecan TFA | 98.7% | 1548.58 | 5 MG
SDP
Catalog No. 5000362251
Encompass_Preferred
Mal-PEG8-Phe-Lys-PAB-Exatecan TFA is an agent-linker conjugate for antibody-drug conjugates (ADC), composed of the degradable linker Mal-PEG8-Phe-Lys-PAB and the toxin molecule Exatecan. This agent is intended for research use only.
  • Composed of degradable linker Mal-PEG8-Phe-Lys-PAB
  • Contains toxin molecule Exatecan
  • Carries a highly potent small-molecule toxin
  • Targets specific antigens of tumor cell surfaces with high binding affinity
  • Internalized into tumor cells and digested in lysosome to release antitumor toxin
Catalog No. 50-003-62251 Supplier Medchemexpress LLC Supplier No. HY148380A5MG
Mal-PEG8-Phe-Lys-PAB-Exatecan TFA is an agent-linker conjugate for antibody-drug conjugates (ADC), composed of the degradable linker Mal-PEG8-Phe-Lys-PAB and the toxin molecule Exatecan. This agent is intended for research use only.
  • Composed of degradable linker Mal-PEG8-Phe-Lys-PAB
  • Contains toxin molecule Exatecan
  • Carries a highly potent small-molecule toxin
  • Targets specific antigens of tumor cell surfaces with high binding affinity
  • Internalized into tumor cells and digested in lysosome to release antitumor toxin