Learn More
Medchemexpress LLC Mal-PEG8-Val-Cit-PAB-MMAF | 100.0% | 1640.95 | C81H129N11O24 | 5 MG
SDP
Catalog No. 5000204393
Encompass_Preferred
Mal-PEG8-Val-Cit-PAB-MMAF is a drug-linker conjugate used in antibody-drug conjugate (ADC) research. It couples the tubulin inhibitor MMAF to a cleavable Val-Cit-PAB peptide linker via an 8-unit PEG spacer and a maleimide reactive group, enabling site-selective conjugation and protease-triggered payload release.
  • Cleavable Val-Cit-PAB peptide linker enables protease-triggered payload release.
  • Maleimide reactive group for cysteine-directed conjugation.
  • PEG8 spacer improves solubility and reduces steric hindrance.
  • High purity suitable for research applications.
  • Molecular weight 1640.95, molecular formula C81H129N11O24.
  • Available in small-mass packs suitable for ADC synthesis and development.
Catalog No. 50-002-04393 Supplier Medchemexpress LLC Supplier No. HY1567585MG
Mal-PEG8-Val-Cit-PAB-MMAF is a drug-linker conjugate used in antibody-drug conjugate (ADC) research. It couples the tubulin inhibitor MMAF to a cleavable Val-Cit-PAB peptide linker via an 8-unit PEG spacer and a maleimide reactive group, enabling site-selective conjugation and protease-triggered payload release.
  • Cleavable Val-Cit-PAB peptide linker enables protease-triggered payload release.
  • Maleimide reactive group for cysteine-directed conjugation.
  • PEG8 spacer improves solubility and reduces steric hindrance.
  • High purity suitable for research applications.
  • Molecular weight 1640.95, molecular formula C81H129N11O24.
  • Available in small-mass packs suitable for ADC synthesis and development.