Learn More
Medchemexpress LLC Mal-PFP ester | 138194-55-5 | 98.2% | 335.18 | 100 MG
SDP
Catalog No. 5000348636
Encompass_Preferred
Mal-PFP ester is an alkyl/ether-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-48636 Supplier Medchemexpress LLC Supplier No. HY138467100MG
Mal-PFP ester is an alkyl/ether-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system
  • Enables selective degradation of target proteins