Learn More
Medchemexpress LLC Mal-PFP ester | 138194-55-5 | 98.2% | 335.18 | 25 MG
SDP
Catalog No. 5000348460
Encompass_Preferred
Mal-PFP ester is an alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-48460 Supplier Medchemexpress LLC Supplier No. 25MGHY138467
Add to Cart
Add to Cart
Mal-PFP ester is an alkyl/ether-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins