Learn More
Medchemexpress LLC Mal-Ph-CONH-PEG4-NHS ester | 1263044-88-7 | 561.54 | 1 G
SDP
Catalog No. 5000365493
Encompass_Preferred
Mal-Ph-CONH-PEG4-NHS ester is a non-cleavable 4 unit PEG ADC linker designed for the synthesis of antibody-drug conjugates (ADCs).
  • Non-cleavable linker
  • 4 unit PEG
  • Used in antibody-drug conjugate synthesis
Catalog No. 50-003-65493 Supplier Medchemexpress LLC Supplier No. 1GHY133545
Mal-Ph-CONH-PEG4-NHS ester is a non-cleavable 4 unit PEG ADC linker designed for the synthesis of antibody-drug conjugates (ADCs).
  • Non-cleavable linker
  • 4 unit PEG
  • Used in antibody-drug conjugate synthesis