Learn More
Medchemexpress LLC Mal-Ph-CONH-PEG4-NHS ester | 1263044-88-7 | ≥97.0% | 561.54 | 500 MG
SDP
Catalog No. 5000366027
Encompass_Preferred
Mal-Ph-CONH-PEG4-NHS ester is an ADC linker with a molecular formula of C26H31N3O11 and a molecular weight of 561.54. This product is related to the Antibody-drug Conjugate/ADC pathway.
  • Non-cleavable 4-unit PEG ADC linker
  • Utilized in the synthesis of antibody-drug conjugates (ADCs)
Catalog No. 50-003-66027 Supplier Medchemexpress LLC Supplier No. 500MGHY133545
Mal-Ph-CONH-PEG4-NHS ester is an ADC linker with a molecular formula of C26H31N3O11 and a molecular weight of 561.54. This product is related to the Antibody-drug Conjugate/ADC pathway.
  • Non-cleavable 4-unit PEG ADC linker
  • Utilized in the synthesis of antibody-drug conjugates (ADCs)