Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-Ph-CONH-PEG4-NHS ester | 1263044-88-7 | >97.0% | 561.54 g/mol | C26H31N3O11 | 100 MG
SDP

Catalog No. 5000202390
Encompass_Preferred

Mal-Ph-CONH-PEG4-NHS ester is a non-cleavable 4-unit polyethylene glycol (PEG4) linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It is designed for use in bioconjugation to prepare antibody-drug conjugates (ADCs), enabling coupling to amine and thiol groups to form stable amide and thioether bonds.

  • Non-cleavable 4-unit PEG linker
  • Maleimide and NHS functional groups for thiol and amine conjugation
  • Suitable for antibody-drug conjugate synthesis and general bioconjugation
  • High purity (≥97.0%)
  • Molecular weight 561.54 g/mol
  • CAS number 1263044-88-7
  • Supplied as a solid in 100 MG packaging

Catalog No. 50-002-02390 Supplier Medchemexpress LLC Supplier No. HY133545100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mal-Ph-CONH-PEG4-NHS ester is a non-cleavable 4-unit polyethylene glycol (PEG4) linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It is designed for use in bioconjugation to prepare antibody-drug conjugates (ADCs), enabling coupling to amine and thiol groups to form stable amide and thioether bonds.

  • Non-cleavable 4-unit PEG linker
  • Maleimide and NHS functional groups for thiol and amine conjugation
  • Suitable for antibody-drug conjugate synthesis and general bioconjugation
  • High purity (≥97.0%)
  • Molecular weight 561.54 g/mol
  • CAS number 1263044-88-7
  • Supplied as a solid in 100 MG packaging

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.