Learn More
Medchemexpress LLC Mal-va-mac-SN38 | 2778229-31-3 | 97.9% | 1040.14 g/mol | C52H61N7O14S | 10 MG
SDP
Catalog No. 5000287948
Encompass_Preferred
Mal-va-mac-SN38 is a drug-linker conjugate for antibody-drug conjugate (ADC) research that delivers the SN-38 cytotoxin via a maleimide-based va-mac linker. It is supplied as a characterized research reagent for in vitro and in vivo ADC studies.
  • High purity (97.9%).
  • Contains SN-38 cytotoxin payload for ADC applications.
  • Maleimide-based va-mac linker for conjugation compatibility.
  • Supplied in a small research quantity (10 MG).
  • Characterized with datasheet, certificate of analysis, and safety data sheet available.
  • Intended for ADC development and cancer research applications.
Catalog No. 50-002-87948 Supplier Medchemexpress LLC Supplier No. HY15956310MG
Mal-va-mac-SN38 is a drug-linker conjugate for antibody-drug conjugate (ADC) research that delivers the SN-38 cytotoxin via a maleimide-based va-mac linker. It is supplied as a characterized research reagent for in vitro and in vivo ADC studies.
  • High purity (97.9%).
  • Contains SN-38 cytotoxin payload for ADC applications.
  • Maleimide-based va-mac linker for conjugation compatibility.
  • Supplied in a small research quantity (10 MG).
  • Characterized with datasheet, certificate of analysis, and safety data sheet available.
  • Intended for ADC development and cancer research applications.