Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-VC-PAB-DM1 | 1464051-44-2 | MFCD10039825 | 99.4% | 1248.81 g/mol | C61H82ClN9O17 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1266825MG

Encompass_Preferred

Mal-VC-PAB-DM1 is a drug-linker conjugate used in the construction of antibody-drug conjugates (ADCs). It links the cytotoxic payload DM1 to a cleavable valine-citrulline-p-aminobenzyl (vc-pab) linker to enable targeted delivery of the payload in ADC research. The compound is supplied as a high-purity material and is soluble in DMSO for research applications.

  • Drug-linker conjugate for ADC research
  • Contains DM1 microtubule-disrupting payload
  • Cleavable valine-citrulline-p-aminobenzyl (vc-pab) linker
  • High purity (99.4%)
  • Molecular weight 1248.81 g/mol
  • Soluble in DMSO
  • CAS number 1464051-44-2

Catalog No. 50-002-02389


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.