Learn More
Medchemexpress LLC Mal-VC-PAB-PNP |10mg
SDP
Catalog No. 5000202552
Encompass_Preferred
Mal-VC-PAB-PNP is a cleavable ADC linker Mal-VC-PAB-PNP can be used in the synthesis of antibody-drug conjugates (ADCs)[1]
Catalog No. 50-002-02552 Supplier Medchemexpress LLC Supplier No. HY14846110MG
Mal-VC-PAB-PNP is a cleavable ADC linker Mal-VC-PAB-PNP can be used in the synthesis of antibody-drug conjugates (ADCs)[1]