Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal3-101 | 831217-40-4 | 98.2% | 931.12 | C54H66N4O10 | 5MG
SDP

Catalog No. 5000239157
Encompass_Preferred

MAL3-101 is a small-molecule allosteric inhibitor of HSP70 used in research to inhibit HSP70 ATPase activity by disrupting Hsp40 co-chaperone interaction. It is supplied as a high-purity solid optimized for in vitro studies and has reliable storage and solubility characteristics for laboratory handling.

  • Allosteric inhibitor of HSP70 that blocks Hsp40 interaction
  • High purity suitable for in vitro research
  • Molecular weight 931.12 and formula C54H66N4O10
  • Off-white to light brown solid appearance
  • Soluble in DMSO (≈50 mg/mL) with ultrasonic assistance
  • Powder stable at -20°C for up to 3 years; in solvent recommended -80°C
  • Applicable to studies of cancer cell proliferation and chaperone biology

Catalog No. 50-002-39157 Supplier Medchemexpress LLC Supplier No. HY1248055MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MAL3-101 is a small-molecule allosteric inhibitor of HSP70 used in research to inhibit HSP70 ATPase activity by disrupting Hsp40 co-chaperone interaction. It is supplied as a high-purity solid optimized for in vitro studies and has reliable storage and solubility characteristics for laboratory handling.

  • Allosteric inhibitor of HSP70 that blocks Hsp40 interaction
  • High purity suitable for in vitro research
  • Molecular weight 931.12 and formula C54H66N4O10
  • Off-white to light brown solid appearance
  • Soluble in DMSO (≈50 mg/mL) with ultrasonic assistance
  • Powder stable at -20°C for up to 3 years; in solvent recommended -80°C
  • Applicable to studies of cancer cell proliferation and chaperone biology

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.