Learn More
Cayman Chemical Malabaricone C, Purity: ≥98%, CAS: 63335-25-1, 10 mg
SDP
Catalog No. 502989475
Encompass_Preferred
A diarylnonanoid with diverse biological activities; scavenges 59.9% of DPPH radicals at 7 μg/ml; active against S. aureus, B. subtilis, and C. albicans in vitro (MICs = 2-32 μg/ml); cytotoxic to A549, HL-60, and MCF-7 cells (IC50s = 12.3, 46.1, and 10.8 μM, respectively); inhibits SMS1 and SMS2 activity in cell lysates (IC50s = 3 and 1.5 μM, respectively); decreases body weight gain, hepatic steatosis, and hepatic and plasma triglyceride levels in a mouse model of high-fat diet-induced obesity at 0.1% in the diet
Catalog No. 50-298-9475 Supplier Cayman Chemical Supplier No. 2974110 MG
A diarylnonanoid with diverse biological activities; scavenges 59.9% of DPPH radicals at 7 μg/ml; active against S. aureus, B. subtilis, and C. albicans in vitro (MICs = 2-32 μg/ml); cytotoxic to A549, HL-60, and MCF-7 cells (IC50s = 12.3, 46.1, and 10.8 μM, respectively); inhibits SMS1 and SMS2 activity in cell lysates (IC50s = 3 and 1.5 μM, respectively); decreases body weight gain, hepatic steatosis, and hepatic and plasma triglyceride levels in a mouse model of high-fat diet-induced obesity at 0.1% in the diet