Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malachite Green 1%, 100 mL, Innovating Science™
GREENER_CHOICESCI_ED

Catalog No. S000989 Shop All Aldon Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000989 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S000989 Supplier Aldon Corporation Supplier No. IS22036
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use 1% aqueous solution of malachite green, suitable for staining applications.

Chemical Identifiers

CAS 7732-18-5 , 569-64-2
Molecular Weight (g/mol) Mixture

Specifications

Color Green
Quantity 100 mL
Percent Purity 1%
Grade Reagent
Chemical Name or Material Malachite green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.