Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malachite Green Oxalate, MP Biomedicals™

Catalog No. MP215264290 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215264290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP215264290 Supplier MP Biomedicals, Inc Supplier No. 0215264290
Add to Cart
Add to Cart
This item is not returnable. View return policy

Malachite Green Oxalate, Green Powder

  • Malachite green is use as counterstain against fuchsin which stains gram-positive and gram-negative bacteria reddish colors and safranin which stains nuclei red.
  • Malachite green stain background to the surrounding tissue blue-green. Malachite green is also used as a pH Indicator between pH 0.2 (green) - 1.8 (blue-green). Used as a botanical counterstain.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2437-29-8 , 33203-82-6
Molecular Formula C52H54N4O12
Molecular Weight (g/mol) 927.02
MDL Number MFCD00011766,MFCD00151209
InChI Key CNYGFPPAGUCRIC-UHFFFAOYSA-L
Synonym Basicgreen 4, Acryl brilliantgreen, Chinagreen, Anilinegreen
IUPAC Name bis(bis[4-(dimethylamino)phenyl](phenyl)methylium) oxalic acid dihydrogen oxalate
SMILES OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 164°C (literature)
UV_Vis 614 - 618 nm
UN Number UN2811
Beilstein 3644550
Color Green
Solubility Information Soluble in water (40mg/ mL), EGME (40mg/ mL), EtOH (50mg/ mL).
Quantity 500 g
Physical Form Powder
Grade Certified Biological Stain
Chemical Name or Material Malachite Green Oxalate
RTECSNumber BQ1190000
storageNote1 Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.