Learn More
Sigma Aldrich Fine Chemicals Biosciences Malachite Green oxalate sa100G
Catalog No. 502898412
Encompass
Malachite Green is a basic dye belonging to the triphenylmethane group. It is required in biology for the staining of microscopic samples such as plant tissue yeast and bacteria. The malachite green method is a sensitive method for the detection of released phosphate.
Catalog No. 50-289-8412 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M9015100G
Malachite Green is a basic dye belonging to the triphenylmethane group. It is required in biology for the staining of microscopic samples such as plant tissue yeast and bacteria. The malachite green method is a sensitive method for the detection of released phosphate.