Learn More
Alkali Scientific Malachite Green, Oxalate Salt, 25 G
SDP
Catalog No. 5000661484
Encompass_Preferred
Malachite Green Oxalate Salt is a dye compound used primarily for laboratory staining purposes. The 25g format is suitable for small-scale laboratory experiments, particularly for visualizing biological samples under a microscope. Malachite green is commonly used to stain fungi, algae, and certain bacterial species in microbiology and histology. Its use is prevalent in studies of cell morphology, metabolic activity, and pathogen detection. The dye interacts with cellular components to produce a vibrant color, enhancing the contrast of specimens during microscopic examination. Malachite Green is often used in academic and clinical laboratories for routine staining procedures.
Catalog No. 50-006-61484 Supplier Alkali Scientific Supplier No. MAL001CP
Malachite Green Oxalate Salt is a dye compound used primarily for laboratory staining purposes. The 25g format is suitable for small-scale laboratory experiments, particularly for visualizing biological samples under a microscope. Malachite green is commonly used to stain fungi, algae, and certain bacterial species in microbiology and histology. Its use is prevalent in studies of cell morphology, metabolic activity, and pathogen detection. The dye interacts with cellular components to produce a vibrant color, enhancing the contrast of specimens during microscopic examination. Malachite Green is often used in academic and clinical laboratories for routine staining procedures.