Learn More
Description
Certified for use as a counterstain for paraffin embedded botanical material; as a spore stain for bacteria by the method of Schaeffer and Fulton.
Specifications
Specifications
| Chemical Name or Material | Malachite Green Oxalate Salt |
| CAS | 2437-29-8 |
| Color | Green |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 463.5 g/mol |
| Percent Purity | ≥90% |
| Grade | Certified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.