Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malachite green oxalate salt, MP Biomedicals™

Catalog No. ICN15264225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15264225 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ICN15264225 Supplier MP Biomedicals, Inc Supplier No. 0215264225
Add to Cart
Add to Cart
This item is not returnable. View return policy

Useful as a stain for bacteria and as a botanical counterstain.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2437-29-8
Molecular Formula C52H54N4O12
Molecular Weight (g/mol) 927.02
MDL Number MFCD00011766,MFCD00151209
InChI Key CNYGFPPAGUCRIC-UHFFFAOYSA-L
Synonym N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate, N, N, N'
PubChem CID 2724411
IUPAC Name bis(bis[4-(dimethylamino)phenyl](phenyl)methylium) oxalic acid dihydrogen oxalate
SMILES OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C

Specifications

Color Green
Quantity 25 g
Physical Form Crystalline Powder
Formula Weight 929
Grade Research
Chemical Name or Material Malachite green oxalate salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.