Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malachite Green Oxalate Salt, Technical Grade, Sigma-Aldrich

Catalog No. 01258881 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-881 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-881 Supplier MilliporeSigma Supelco Supplier No. M6880100G
Add to Cart
Add to Cart

Technical grade reagent used in diagnostic assays, hematology, and histology for phosphate detection.

Malachite Green oxalate salt has been used in biliary phospholipid analysis.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Malachite Green Oxalate Salt
CAS 2437-29-8
Color Green
Quantity 100 g
Molecular Weight (g/mol) 463.5 g/mol
Grade Technical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.