Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malachite Green (Technical), Fisher Chemical

Catalog No. A779500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A779-500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A779-500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 569-64-2
Molecular Formula C23H25ClN2
Molecular Weight (g/mol) 364.92
MDL Number MFCD00011751
InChI Key FDZZZRQASAIRJF-UHFFFAOYSA-M
Synonym China Green, Basic Green 4, Fast Green
PubChem CID 11294
ChEBI CHEBI:72449
IUPAC Name bis[4-(dimethylamino)phenyl](phenyl)methylium chloride
SMILES [Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C

Specifications

Identification Pass Test
Formula Weight 364.9
Grade Technical
Packaging Poly Bottle
Color Index No. 42000
Color Brown
Quantity 500 g
Physical Form Solid
Chemical Name or Material Malachite Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.