Learn More
Cayman Chemical Malachite Green (chloride), Purity: ≥95%, CAS: 569-64-2, 5 g
SDP
Catalog No. 502918172
Encompass_Preferred
A triphenylmethane dye; inhibits the proliferation of HEp-2 human larynx cells (IC50 = 2.06 µM); cytotoxic to HEp-2 cells and Caco-2 human colorectal adenocarcinoma cells (IC50s = 2.03 and 13.8 µM, respectively); reduces fungal infection and increases hatching rates of rainbow trout eggs at 2 µg/ml for 60 minutes twice a week; used as a biochemical indicator dye for titrations, pH indicators, and counterstains in histology applications
Catalog No. 50-291-8172 Supplier Cayman Chemical Supplier No. 373955 G
A triphenylmethane dye; inhibits the proliferation of HEp-2 human larynx cells (IC50 = 2.06 µM); cytotoxic to HEp-2 cells and Caco-2 human colorectal adenocarcinoma cells (IC50s = 2.03 and 13.8 µM, respectively); reduces fungal infection and increases hatching rates of rainbow trout eggs at 2 µg/ml for 60 minutes twice a week; used as a biochemical indicator dye for titrations, pH indicators, and counterstains in histology applications