Learn More
Selleck Chemical LLC Malantide P1156-5MG
SDP
Catalog No. 5000041547
Encompass_Preferred
Malantide is a synthetic dodecapeptide derived from the site phosphorylated by cAMP-dependent protein kinase (PKA) on the -subunit of phosphorylase kinase which is a highly specific substrate for PKA with a Km of 15 M
Catalog No. 50-000-41547 Supplier Selleck Chemical LLC Supplier No. P11565MG
Malantide is a synthetic dodecapeptide derived from the site phosphorylated by cAMP-dependent protein kinase (PKA) on the -subunit of phosphorylase kinase which is a highly specific substrate for PKA with a Km of 15 M