Learn More
Bioworld Malaria Protein HSP (Recombinant), 100 μg
Supplier:  Bioworld 220605481
Encompass_Preferred
Recombinant Malaria Protein HSP (MAL) Source: E.Coli Background: Description: E.coli derived recombinant. The protein contains the Malaria HSP protein epitopes (33-114a.a). Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. Protein concentration: 1 mg/ml containing 150mM Imidazole, pH 8.0, 150mM NaCl, 150mM Sodium Phosphate and 50% glycerol. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -200C. Stability: Five years frozen. One month in solution at room temperature. Application: Use as an antigen in ELISA and Western Blots. Usage: BioWORLD\\\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-648