Learn More
ATCC Azotobacter chroococcum
Supplier:  ATCC 4412
Encompass
Azotobacter chroococcum strain X-50 is a whole-genome sequenced bacterium that was isolated from soil. This culture has potential applications as a plant growth promoting strain.
Catalog No. 50-238-1109
 Restricted Item