Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MALAT1-IN-1 50mg | 827327-28-6 | 50 MG
SDP

Catalog No. 5000228938
Encompass_Preferred

MALAT1-IN-1 is a small-molecule inhibitor of the MALAT1 long non-coding RNA supplied as a solid research reagent for in vitro studies of MALAT1-mediated pathways and cancer biology. The compound is provided at 50 mg and is characterized by high purity, defined storage conditions, and DMSO solubility suitable for laboratory assays.

  • High purity (99.8%).
  • Molecular formula C19H21N3O2, molecular weight 323.39 g/mol.
  • Appearance: white to light yellow solid.
  • Soluble in DMSO (100 mg/mL); ultrasonic agitation may be needed.
  • Recommended storage: powder -20 °C (long term) or 4 °C; in solvent -80 °C or -20 °C.
  • Supplied as a 50 mg solid and as a 10 mM solution in DMSO.

Catalog No. 50-002-28938 Supplier Medchemexpress LLC Supplier No. HY11557950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MALAT1-IN-1 is a small-molecule inhibitor of the MALAT1 long non-coding RNA supplied as a solid research reagent for in vitro studies of MALAT1-mediated pathways and cancer biology. The compound is provided at 50 mg and is characterized by high purity, defined storage conditions, and DMSO solubility suitable for laboratory assays.

  • High purity (99.8%).
  • Molecular formula C19H21N3O2, molecular weight 323.39 g/mol.
  • Appearance: white to light yellow solid.
  • Soluble in DMSO (100 mg/mL); ultrasonic agitation may be needed.
  • Recommended storage: powder -20 °C (long term) or 4 °C; in solvent -80 °C or -20 °C.
  • Supplied as a 50 mg solid and as a 10 mM solution in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.