Learn More
Medchemexpress LLC HY-115579 5mg Medchemexpress, MALAT1-IN-1 CAS:827327-28-6 Purity:>98%
SDP
Catalog No. 501969397
Encompass_Preferred
Medchemexpress, HY-115579 5mg MALAT1-IN-1 CAS:827327-28-6 MALAT1-IN-1 (compounds 5) is a potent and specific Malat1 (Metastasis-associated lung adenocarcinoma transcript 1) inhibitor. MALAT1-IN-1 modulated Malat1 downstream genes in a dose-dependent manner without affecting expression of nuclear enriched abundant transcript 1 (Neat1) . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9397 Supplier Medchemexpress LLC Supplier No. HY1155795MG
Medchemexpress, HY-115579 5mg MALAT1-IN-1 CAS:827327-28-6 MALAT1-IN-1 (compounds 5) is a potent and specific Malat1 (Metastasis-associated lung adenocarcinoma transcript 1) inhibitor. MALAT1-IN-1 modulated Malat1 downstream genes in a dose-dependent manner without affecting expression of nuclear enriched abundant transcript 1 (Neat1) . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.