Learn More
Cayman Chemical Malate Dehydrogenase 1humn
SDP
Catalog No. 502943250
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged MDH1 expressed in E. coli • Amino acids: 1-337 • MW: 37.5 kDa
Catalog No. 50-294-3250 Supplier Cayman Chemical Supplier No. 40110100 UG
Source: Active recombinant human C-terminal His-tagged MDH1 expressed in E. coli • Amino acids: 1-337 • MW: 37.5 kDa