Learn More
Bioworld Malate Dehydrogenase, 2.5 KU
Catalog No. 50198985
Encompass_Preferred
Malate DehydrogenaseSpecification:Malate Dehydrogenase, Ammonium Sulfate SuspensionUltra Pure GradeSource: Porcine heartSpecific activity (25 °C): > 1200 U/mgConcentration: > 12,000 U/mLAlanine aminotransferase: < 0.005%Aspartate aminotransferase: < 0.005%Lactate dehydrogenase: < 0.005%Unit Definition: The amount of enzyme which will convert 1 micromole of oxaloacetate and NADH per minute at 25 C in phosphate buffer at pH 7.5.
Catalog No. 50-198-985 Supplier Bioworld Supplier No. 215600181
Malate DehydrogenaseSpecification:Malate Dehydrogenase, Ammonium Sulfate SuspensionUltra Pure GradeSource: Porcine heartSpecific activity (25 °C): > 1200 U/mgConcentration: > 12,000 U/mLAlanine aminotransferase: < 0.005%Aspartate aminotransferase: < 0.005%Lactate dehydrogenase: < 0.005%Unit Definition: The amount of enzyme which will convert 1 micromole of oxaloacetate and NADH per minute at 25 C in phosphate buffer at pH 7.5.