Learn More
Novus Biologicals™ Malate Dehydrogenase Activity Assay Kit (Fluorometric)
Description
In this assay, MDH converts malate into intermediate products, which will further react with the Probe forming a fluorophore. Detection method: Fluorescence (Ex/Em = 535/587nm). The kit is simple, sensitive and high-throughput adaptable and can detect as low as 40 uU of MDH activity.
- Suitable for the analysis of citric acid cycle and malate-asparate shuttle
- Can be used with animal heart, liver, muscle tissues
- Can be used with purified mitochondria
- Can be used with adherent or suspension cell cultures
Kit Contents:
- Probe
- NADH Standard
- MDH Assay Buffer
- MDH Developer
- MDH Positive Control
- MDH Substrate
Specifications
Specifications
| Quantity | 100 Assays |
| Product Type | Assay Kit (Fluorometric) |
| Content And Storage | Store at -20°C in the dark. Avoid freeze-thaw cycles. |