Learn More
Chem Service Inc Malathion, 121-75-5, MFCD00036233, 100 mg
SDP
Catalog No. NC1440276
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula: C10-H19-O6-P-S2, Molecular Weight: 330.36, Classification: Insecticide, Acaricide, Acaricide. Malathion is a commonly used organophosphate pesticide, which is generally sprayed on aquatic habitats in order to have a control over mosquitoes that carry malaria and the west nile virus.
Catalog No. NC1440276 Supplier Chem Service Inc Supplier No. N12346100MG
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula: C10-H19-O6-P-S2, Molecular Weight: 330.36, Classification: Insecticide, Acaricide, Acaricide. Malathion is a commonly used organophosphate pesticide, which is generally sprayed on aquatic habitats in order to have a control over mosquitoes that carry malaria and the west nile virus.