Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Malathion, >95%, Ultra Scientific

Catalog No. USPST641
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PST-641 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PST-641 Supplier Agilent Technologies Supplier No. PST641
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 121-75-5
Molecular Formula C10H19O6PS2
Molecular Weight (g/mol) 330.35
InChI Key JXSJBGJIGXNWCI-UHFFFAOYSA-N
PubChem CID 4004
ChEBI CHEBI:6651
IUPAC Name diethyl 2-dimethoxyphosphinothioylsulfanylbutanedioate
SMILES CCOC(=O)CC(C(=O)OCC)SP(=S)(OC)OC

Specifications

CAS Max % 100.0
Assay Percent Range >95 % min.
Solvent or Matrix Neat
Quantity 100 mg
Chemical Name or Material Malathion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.