Learn More
Crescent Chemical Co Inc MALATHION SOLN 1ML
SDP
Supplier:  Crescent Chemical Co Inc CT7717M.10
Encompass_Preferred
Malathion Single Component Calibration Standard in Methanol CAS 121-75-5 Size - 1mL Catalog Number - 7717M.10
Catalog No. 50-711-05