Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MALDI Validation Set, Polystyrene Mp ∼700 - 70,000, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111002903 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2903 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2903 Supplier MilliporeSigma Supelco Supplier No. 035651EA
Add to Cart
Add to Cart

Specifications

Description MALDI Standards are suitable for the validation and comparison of MALDI-ToF instruments. Certificates provide MALDI spectra including Mw, Mn and Mp values of select narrow polymers, which differ in molar mass. This set inclces 6 different polystyrene standards ranging from Mp ˜700 to ˜70,000.Each standard is characterized with GPC, a second absolute method (light scattering or H-NMR) and MALDI-ToF-MS.
Linear Formula [CH2CH(C6H5)]n
MDL Number MFCD00084450
Synonym Polystyrene; PS
Quantity 1 Ea.
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.