Learn More
New Jersey Fluid Systems LLC MALE ADAPT 1/4 TUBE TO MLE NPT
Catalog No. NC9422985
Encompass
316 Stainless Steel Swagelok Tube Fitting Adapter. !/4 in. OD Tube Stub x 1/4 in. Male NPT
Catalog No. NC9422985 Supplier New Jersey Fluid Systems LLC Supplier No. SS4TA14
316 Stainless Steel Swagelok Tube Fitting Adapter. !/4 in. OD Tube Stub x 1/4 in. Male NPT