Learn More
New Jersey Fluid Systems LLC MALE CONNECTOR 1/8 INCH
Catalog No. NC9334095
Encompass
316 Stainless Steel Swagelok Tube Fitting Male Connector. 1/8 in. OD Tube Fitting x 1/8 in. Male NPT
Catalog No. NC9334095 Supplier New Jersey Fluid Systems LLC Supplier No. SS20012
316 Stainless Steel Swagelok Tube Fitting Male Connector. 1/8 in. OD Tube Fitting x 1/8 in. Male NPT