Learn More
New Jersey Fluid Systems LLC MALE CONNECTOR
Supplier:  New Jersey Fluid Systems LLC SS60014
Encompass
316 Stainless Steel Swagelok Tube Fitting Male Connector. 3/8 in. OD x 1/4 in. Male NPT
Catalog No. NC9318032