Learn More
COLDER PRODUCTS COMPANY MALE COUPLING 1/4 25/PK
Catalog No. 503131450
Encompass_Preferred
CPC Male Delrin Coupling Connector Gray with Dark Gray Overmold For Use With 6 4mm Includes Shut-off valves EPDM Seal Overall Length 1 98 in 50 mm Hose Barb Valved In-line Coupling Insert Height 0 96 in 24 3 mm Maximum Pressure 120 psi
Catalog No. 50-313-1450 Supplier COLDER PRODUCTS COMPANY Supplier No. NS4D22004
CPC Male Delrin Coupling Connector Gray with Dark Gray Overmold For Use With 6 4mm Includes Shut-off valves EPDM Seal Overall Length 1 98 in 50 mm Hose Barb Valved In-line Coupling Insert Height 0 96 in 24 3 mm Maximum Pressure 120 psi