Learn More
COLDER PRODUCTS COMPANY MALE COUPLING 1/4 25/PK
Catalog No. 503131324
Encompass_Preferred
CPC Colder Products Company NS4 Series Quick-Disconnect Coupling Bodies and Inserts Overall Length 49 5 mm 1 95 in Gray with Dark Gray Overmold For Use With 6 4mm Includes Shut-off valves Polypropylene NPT Valved Coupling Insert Height 0 96 in 24 3 mm Maximum Pressure 120 psi
Catalog No. 50-313-1324 Supplier COLDER PRODUCTS COMPANY Supplier No. NS4D24004
CPC Colder Products Company NS4 Series Quick-Disconnect Coupling Bodies and Inserts Overall Length 49 5 mm 1 95 in Gray with Dark Gray Overmold For Use With 6 4mm Includes Shut-off valves Polypropylene NPT Valved Coupling Insert Height 0 96 in 24 3 mm Maximum Pressure 120 psi