Learn More
COLDER PRODUCTS COMPANY MALE COUPLING 1/8 25/PK
Catalog No. 503131323
Encompass_Preferred
CPC Colder Products Company NS4 Series Quick-Disconnect Coupling Bodies and Inserts Overall Length 44 mm 1 73 in Gray with Dark Gray Overmold For Use With 3 2mm Includes Shut-off valves EPDM Seal Hose Barb Valved In-line Coupling Insert Height 0 96 in 24 3 mm
Catalog No. 50-313-1323 Supplier COLDER PRODUCTS COMPANY Supplier No. NS4D22002
CPC Colder Products Company NS4 Series Quick-Disconnect Coupling Bodies and Inserts Overall Length 44 mm 1 73 in Gray with Dark Gray Overmold For Use With 3 2mm Includes Shut-off valves EPDM Seal Hose Barb Valved In-line Coupling Insert Height 0 96 in 24 3 mm