Learn More
COLDER PRODUCTS COMPANY CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Insert Polycarbonate w/Silicone Seal USP Class VI Diameter Inner 9 5 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Purple
Catalog No. 503131445
Encompass_Preferred
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Insert Polycarbonate w/Silicone Seal USP Class VI Diameter Inner 9 5 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Purple
This item is not returnable. View return policy
Catalog No. 50-313-1445 Supplier COLDER PRODUCTS COMPANY Supplier No. MPC22006T03M
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Insert Polycarbonate w/Silicone Seal USP Class VI Diameter Inner 9 5 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Purple